Assay Details
Gene Symbol: KRTDAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244847.1 | NP_001231776.1 | 4 - 5 | 310 | 65 | |
NM_207392.2 | NP_997275.1 | 5 - 6 | 352 | 65 | ||
GenBank mRNA | AY359007.1 | - | 5 - 6 | 275 | 65 | |
BC130501.1 | - | 5 - 6 | 352 | 40146532 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
