Assay Details
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139242.3 | NP_640335.2 | 4 - 5 | 672 | 85 | |
XM_005254158.4 | XP_005254215.2 | 4 - 5 | 814 | 85 | ||
GenBank mRNA | BC016630.1 | - | 4 - 5 | 650 | 4707448 | 85 |
BC033687.1 | - | 4 - 5 | 660 | 5196677 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001557.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15014834.hg.1 | ENST00000220058.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014834.hg.1 | ENST00000560717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014834.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15014834.hg.1 | TC15001557.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172221.hg.1 | TC1500009773.hg.1 | Clariom D TC |
