Assay Details
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139242.3 | NP_640335.2 | 5 - 6 | 750 | 90 | |
XM_005254158.4 | XP_005254215.2 | 5 - 6 | 892 | 90 | ||
GenBank mRNA | AK055688.1 | - | 5 - 6 | 696 | 90 | |
AK301390.1 | - | 3 - 4 | 524 | 90 | ||
BC016630.1 | - | 5 - 6 | 728 | 4707448 | 90 | |
BC033687.1 | - | 5 - 6 | 738 | 5196677 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001557.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15014833.hg.1 | ENST00000220058.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014833.hg.1 | ENST00000560717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014833.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15014833.hg.1 | TC15001557.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500172220.hg.1 | TC1500009773.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000220058.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000543678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558460.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000558614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000559633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000560717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | ENST00000561025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009773.hg.1 | NM_139242.3 | Refseq |
