Assay Details
| Gene Symbol: KIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012311.3 | NP_036443.1 | 6 - 7 | 701 | 100 | |
| NR_045609.1 | 6 - 7 | 701 | 100 | |||
| XM_006717434.3 | XP_006717497.1 | 6 - 7 | 696 | 100 | ||
| XM_006717435.3 | XP_006717498.1 | 5 - 6 | 569 | 100 | ||
| XM_011519427.1 | XP_011517729.1 | 6 - 7 | 696 | 100 | ||
| XM_017015946.1 | XP_016871435.1 | 5 - 6 | 572 | 100 | ||
| GenBank mRNA | AJ005273.1 | - | 6 - 7 | 672 | 100 | |
| AK296637.1 | - | 5 - 6 | 569 | 100 | ||
| AK301789.1 | - | 6 - 7 | 701 | 100 | ||
| AK309059.1 | - | 6 - 7 | 637 | 100 | ||
| BC017309.2 | - | 6 - 7 | 618 | 5089578 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001022.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | ENST00000379562.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | ENST00000535925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | ENST00000543003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012740.hg.1 | TC10001022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000166530.hg.1 | TC1000009742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |

KIN
Genomic Map