Assay Details
Gene Symbol: PADI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007365.2 | NP_031391.2 | 14 - 15 | 62 | ||
XM_017000148.1 | XP_016855637.1 | 6 - 7 | 62 | |||
GenBank mRNA | AB023211.1 | - | 14 - 15 | 62 | ||
AB030176.1 | - | 14 - 15 | 1697 | 62 | ||
AK127006.1 | - | 5 - 6 | 1989 | 62 | ||
AK295783.1 | - | 11 - 12 | 1367 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002287.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01034858.hg.1 | TC01002287.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01034858.hg.1 | ENST00000375486.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034858.hg.1 | ENST00000444885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034858.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034858.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196646.hg.1 | TC0100013076.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
