Assay Details
Gene Symbol: PADI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007365.2 | NP_031391.2 | 7 - 8 | 919 | 60 | |
GenBank mRNA | AB023211.1 | - | 7 - 8 | 901 | 60 | |
AB030176.1 | - | 7 - 8 | 901 | 60 | ||
BC009701.1 | - | 7 - 8 | 901 | 3901790 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002287.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01034870.hg.1 | TC01002287.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01034870.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034870.hg.1 | ENST00000375486.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01034870.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196658.hg.1 | TC0100013076.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
