Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004401.2 | NP_004392.1 | 1 - 2 | 230 | 61 | |
NM_213566.1 | NP_998731.1 | 1 - 2 | 230 | 61 | ||
GenBank mRNA | AF087573.1 | - | 1 - 2 | 169 | 61 | |
AF103799.1 | - | 1 - 2 | 157 | 61 | ||
AK222546.1 | - | 1 - 2 | 166 | 61 | ||
AU121791.1 | - | 1 - 2 | 230 | 61 | ||
BC000037.2 | - | 1 - 2 | 150 | 3505817 | 61 | |
BC007112.1 | - | 1 - 2 | 201 | 4292811 | 61 | |
BC007721.2 | - | 1 - 2 | 162 | 4299734 | 61 | |
BT006980.1 | - | 1 - 2 | 132 | 61 | ||
U91985.1 | - | 1 - 2 | 188 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002190.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01033733.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033733.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033733.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033729.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033729.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033729.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033729.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033729.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033728.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033728.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033728.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033728.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033728.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033732.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033732.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033732.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033732.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006477.hg.1 | ENST00000517626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006477.hg.1 | ENST00000517626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194842.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006477.hg.1 | ENST00000517626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006477.hg.1 | ENST00000517626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194848.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194848.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194848.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194848.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194848.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194843.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194847.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 137503 | - | - |
