Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004401.2 | NP_004392.1 | 5 - 6 | 880 | 94 | |
GenBank mRNA | AK222546.1 | - | 5 - 6 | 816 | 94 | |
AK313317.1 | - | 5 - 6 | 814 | 94 | ||
BC000037.2 | - | 5 - 6 | 800 | 3505817 | 94 | |
BC007112.1 | - | 5 - 6 | 851 | 4292811 | 94 | |
BC007721.2 | - | 5 - 6 | 812 | 4299734 | 94 | |
BT006980.1 | - | 5 - 6 | 782 | 94 | ||
U91985.1 | - | 5 - 6 | 838 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002190.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01033718.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033718.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033718.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033718.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033720.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033720.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033720.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033720.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033715.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033715.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033715.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033715.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033719.hg.1 | TC01002190.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033719.hg.1 | ENST00000377038.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033719.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033719.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194829.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194832.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377036.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012846.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194830.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194831.hg.1 | TC0100012846.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | ENST00000377038.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | ENST00000476658.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | NM_004401.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | NM_213566.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194833.hg.1 | TC0100012846.hg.1 | Clariom D TC |
