Assay Details
| Gene Symbol: TRAPPC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079537.1 | NP_001073005.1 | 4 - 5 | 681 | 77 | |
| XM_011536409.2 | XP_011534711.1 | 5 - 6 | 701 | 77 | ||
| XM_017020964.1 | XP_016876453.1 | 4 - 5 | 654 | 77 | ||
| GenBank mRNA | AK294339.1 | - | 2 - 3 | 379 | 77 | |
| AK310730.1 | - | 4 - 5 | 491 | 77 | ||
| AK315900.1 | - | 4 - 5 | 609 | 77 | ||
| AL833179.1 | - | 4 - 5 | 565 | 77 | ||
| BX161510.1 | - | 4 - 5 | 573 | 77 | ||
| BX248013.1 | - | 4 - 5 | 637 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001064.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | ENST00000330149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | ENST00000347691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | ENST00000557764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012122.hg.1 | TC14001064.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166298.hg.1 | TC1400009006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000330149.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000347691.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000469361.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000554018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000555269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000556765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | ENST00000557764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_001079537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009006.hg.1 | NM_177452.3 | Refseq |

TRAPPC6B
Genomic Map