Assay Details
| Gene Symbol: ABCG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022436.2 | NP_071881.1 | 2 - 3 | 404 | 57 | |
| XM_005264480.3 | XP_005264537.1 | 2 - 3 | 425 | 57 | ||
| XM_006712073.3 | XP_006712136.1 | 2 - 3 | 425 | 57 | ||
| XM_006712074.3 | XP_006712137.1 | 2 - 3 | 425 | 57 | ||
| XM_011533024.2 | XP_011531326.1 | 2 - 3 | 425 | 57 | ||
| XM_011533025.2 | XP_011531327.1 | 2 - 3 | 849 | 57 | ||
| XM_011533026.2 | XP_011531328.1 | 2 - 3 | 425 | 57 | ||
| GenBank mRNA | AF312715.2 | - | 2 - 3 | 404 | 57 | |
| AF320293.1 | - | 2 - 3 | 370 | 57 | ||
| AK091997.1 | - | 1 - 2 | 1031 | 57 | ||
| BC111541.1 | - | 1 - 2 | 916 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111541.1, XM_011533025.2 transcripts.
Set Membership: |
|

ABCG5
Genomic Map