Assay Details
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145664.1 | NP_001139136.1 | 4 - 5 | 361 | 93 | |
XM_011511771.2 | XP_011510073.1 | 5 - 6 | 899 | 93 | ||
XM_011511772.1 | XP_011510074.1 | 7 - 8 | 573 | 93 | ||
XM_011511773.1 | XP_011510075.1 | 3 - 4 | 244 | 93 | ||
XM_017004851.1 | XP_016860340.1 | 7 - 8 | 583 | 93 | ||
XM_017004852.1 | XP_016860341.1 | 4 - 5 | 370 | 93 | ||
XM_017004853.1 | XP_016860342.1 | 7 - 8 | 583 | 93 | ||
XM_017004854.1 | XP_016860343.1 | 7 - 8 | 583 | 93 | ||
GenBank mRNA | AK298610.1 | - | 4 - 5 | 361 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02005042.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02035097.hg.1 | TC02005042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02035097.hg.1 | ENST00000376826.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035097.hg.1 | ENST00000428343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035097.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02035104.hg.1 | TC02005042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02035104.hg.1 | ENST00000428343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035104.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02035092.hg.1 | TC02005042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02035092.hg.1 | ENST00000376826.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035092.hg.1 | ENST00000428343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035092.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02035102.hg.1 | TC02005042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02035102.hg.1 | ENST00000376826.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035102.hg.1 | ENST00000428343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035102.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02035096.hg.1 | TC02005042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02035096.hg.1 | ENST00000376826.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035096.hg.1 | ENST00000428343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02035096.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200350.hg.1 | TC0200013669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200343.hg.1 | TC0200013669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200357.hg.1 | TC0200013669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200348.hg.1 | TC0200013669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200356.hg.1 | TC0200013669.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | ENST00000492238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013669.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000428343.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000481179.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | ENST00000484091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | NM_001145664.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200200355.hg.1 | TC0200013669.hg.1 | Clariom D TC |
