Assay Details
| Gene Symbol: TMEM185B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024121.2 | NP_077026.2 | 1 | 5136 | 107 | |
| GenBank mRNA | AK090980.1 | - | 1 | 1242 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437910 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437924 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552849 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437933 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552865 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15315941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21552871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01437890 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3594654 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P119789 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P119789 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15803675 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15803675 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3314476 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3314476 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 219253_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 219253_at | - | - |
| Human Genome U133 Plus 2.0 Array | 219253_PM_at | - | - |
| Human Genome U133A 2.0 Array | 219253_PM_at | - | - |
| Human Genome U133A 2.0 Array | 219253_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw550787 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 219253_at7 | - | - |
| Human Genome U95 Set | 45603_PM_at | - | - |
| Human Genome U95 Set | 45603_at | - | - |
| Human Genome U133 Set | 219253_at | - | - |
| Human Genome U133 Set | 219253_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014265 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014267 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014266 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014267 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014265 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 137290 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 707372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2231021 | BC080607.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231021 | CR595954.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231021 | CR626230.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231021 | AF530474.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231021 | NM_024121.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231020 | BC080607.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231020 | CR595954.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231020 | CR626230.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231020 | AF530474.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231020 | NM_024121.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1696996 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231021 | BC080607.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231021 | CR595954.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231021 | CR626230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231021 | AF530474.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231021 | NM_024121.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231020 | BC080607.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231020 | CR595954.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231020 | CR626230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231020 | AF530474.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231020 | NM_024121.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1696996 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231020 | BC080607.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231020 | CR595954.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231020 | CR626230.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231020 | AF530474.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231020 | NM_024121.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231021 | BC080607.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231021 | CR595954.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231021 | CR626230.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231021 | AF530474.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231021 | NM_024121.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231020 | BC080607.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231020 | CR595954.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231020 | CR626230.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231020 | AF530474.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231020 | NM_024121.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231020 | BC080607.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231020 | CR595954.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231020 | CR626230.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231020 | AF530474.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231020 | NM_024121.2 | human_transcriptome |

TMEM185B
Genomic Map