Assay Details
| Gene Symbol: SSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256732.1 | NP_001243661.1 | 2 - 3 | 344 | 153 | |
| NM_001256733.1 | NP_001243662.1 | 2 - 3 | 344 | 153 | ||
| NM_001256734.1 | NP_001243663.1 | 2 - 3 | 344 | 153 | ||
| NM_001256735.1 | NP_001243664.1 | 2 - 3 | 344 | 153 | ||
| NM_001256736.1 | NP_001243665.1 | 2 - 3 | 344 | 153 | ||
| NM_012446.3 | NP_036578.2 | 2 - 3 | 344 | 153 | ||
| XM_011543297.2 | XP_011541599.1 | 2 - 3 | 259 | 153 | ||
| XM_017009306.1 | XP_016864795.1 | 2 - 3 | 259 | 153 | ||
| GenBank mRNA | AF077048.1 | - | 2 - 3 | 181 | 153 | |
| AF161465.1 | - | 2 - 3 | 175 | 152 | ||
| AK074537.1 | - | 2 - 3 | 139 | 153 | ||
| AK293544.1 | - | 2 - 3 | 139 | 153 | ||
| AK294864.1 | - | 2 - 3 | 139 | 153 | ||
| AK297494.1 | - | 2 - 3 | 139 | 153 | ||
| AK299858.1 | - | 2 - 3 | 188 | 153 | ||
| AK312337.1 | - | 2 - 3 | 188 | 153 | ||
| AL080076.1 | - | 2 - 3 | 192 | 153 | ||
| BC017020.1 | - | 2 - 3 | 192 | 3913337 | 153 | |
| BI517313.1 | - | 2 - 3 | 344 | 153 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF161465.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SSBP2
Genomic Map