Assay Details
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152751.2 | NP_689964.2 | 3 - 4 | 590 | 92 | |
XM_005252402.4 | XP_005252459.1 | 3 - 4 | 688 | 92 | ||
XM_011519392.2 | XP_011517694.1 | 3 - 4 | 687 | 92 | ||
XM_017015918.1 | XP_016871407.1 | 3 - 4 | 622 | 92 | ||
XM_017015919.1 | XP_016871408.1 | 3 - 4 | 644 | 92 | ||
GenBank mRNA | AK097602.1 | - | 3 - 4 | 590 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001051.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | ENST00000341083.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | ENST00000396898.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | ENST00000396900.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10013068.hg.1 | TC10001051.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186656.hg.1 | TC1000012522.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000341083.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000378605.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000440282.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000463303.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000469555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000480703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000486542.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | ENST00000492675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_001100912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012522.hg.1 | NM_152751.2 | Refseq |
