Assay Details
Gene Symbol: PRNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000311.3 | NP_000302.1 | 1 - 2 | 362 | 95 | |
NM_001271561.1 | NP_001258490.1 | 1 - 2 | 362 | 95 | ||
GenBank mRNA | BG397054.1 | - | 91 | 95 | ||
DA122620.1 | - | 1 - 2 | 302 | 95 | ||
DA297032.1 | - | 1 - 2 | 333 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_000311.3, NM_001271561.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
cellular copper ion homeostasis
response to oxidative stress
cell cycle arrest
learning or memory
metabolic process
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
negative regulation of interleukin-2 production
regulation of protein localization
cellular response to drug
negative regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of activated T cell proliferation
response to cadmium ion
negative regulation of T cell receptor signaling pathway
protein homooligomerization
negative regulation of calcineurin-NFAT signaling cascade
cellular response to copper ion
regulation of potassium ion transmembrane transport
copper ion binding
protein binding
lamin binding
microtubule binding
tubulin binding
identical protein binding
ATP-dependent protein binding
ion channel binding
chaperone binding
negative regulation of protein phosphorylation
cellular copper ion homeostasis
response to oxidative stress
cell cycle arrest
learning or memory
metabolic process
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
negative regulation of interleukin-2 production
regulation of protein localization
cellular response to drug
negative regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of activated T cell proliferation
response to cadmium ion
negative regulation of T cell receptor signaling pathway
protein homooligomerization
negative regulation of calcineurin-NFAT signaling cascade
cellular response to copper ion
regulation of potassium ion transmembrane transport
copper ion binding
protein binding
lamin binding
microtubule binding
tubulin binding
identical protein binding
ATP-dependent protein binding
ion channel binding
chaperone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000379440.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000424424.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000430350.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | ENST00000457586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_000311.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080122.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001080123.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_001271561.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006576.hg.1 | NM_183079.3 | Refseq |
