Assay Details
| Gene Symbol: PLA2G4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206670.1 | NP_001193599.1 | 12 - 13 | 1252 | 75 | |
| XM_011521237.2 | XP_011519539.1 | 12 - 13 | 1162 | 75 | ||
| XM_011521238.1 | XP_011519540.1 | 3 - 4 | 970 | 75 | ||
| GenBank mRNA | AK127558.1 | - | 2 - 3 | 541 | 75 | |
| BC101584.1 | - | 1 - 2 | 157 | 75 | ||
| BC101612.1 | - | 1 - 2 | 157 | 75 | ||
| BX648482.1 | - | 2 - 3 | 451 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001258.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid metabolic process
phospholipid catabolic process
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
phospholipid metabolic process
phospholipid catabolic process
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15011495.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011495.hg.1 | ENST00000413860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011495.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011495.hg.1 | TC15001258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166540.hg.1 | TC1500009177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |

PLA2G4E
Genomic Map