Assay Details
| Gene Symbol: PRSS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022119.3 | NP_071402.1 | 2 - 3 | 169 | 81 | |
| XM_005255473.3 | XP_005255530.1 | 3 - 4 | 268 | 81 | ||
| GenBank mRNA | AB010779.1 | - | 2 - 3 | 106 | 81 | |
| AF321182.1 | - | 2 - 3 | 120 | 81 | ||
| AY358396.1 | - | 2 - 3 | 155 | 81 | ||
| BC009726.1 | - | 2 - 3 | 141 | 3899480 | 81 | |
| BQ893064.1 | - | 177 | 81 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000795.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011639.hg.1 | ENST00000161006.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011639.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011639.hg.1 | TC16000795.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169671.hg.1 | TC1600009147.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |

PRSS22
Genomic Map