Assay Details
Gene Symbol: PRSS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022119.3 | NP_071402.1 | 3 - 4 | 343 | 69 | |
XM_005255473.3 | XP_005255530.1 | 4 - 5 | 442 | 69 | ||
GenBank mRNA | AB010779.1 | - | 3 - 4 | 280 | 69 | |
AF321182.1 | - | 3 - 4 | 294 | 69 | ||
AK309624.1 | - | 3 - 4 | 485 | 69 | ||
AY358396.1 | - | 3 - 4 | 329 | 69 | ||
BC009726.1 | - | 3 - 4 | 315 | 3899480 | 69 | |
BQ893064.1 | - | 351 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000795.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16011638.hg.1 | ENST00000161006.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16011638.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16011638.hg.1 | TC16000795.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000571228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009147.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169670.hg.1 | TC1600009147.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169665.hg.1 | TC1600009147.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000161006.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000570950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000572061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000574768.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000575164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000576381.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | ENST00000577177.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | NM_022119.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600169666.hg.1 | TC1600009147.hg.1 | Clariom D TC |
