Assay Details
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014043.3 | NP_054762.2 | 2 - 3 | 381 | 98 | |
XM_011533576.2 | XP_011531878.1 | 2 - 3 | 212 | 98 | ||
GenBank mRNA | AF151842.1 | - | 2 - 3 | 340 | 98 | |
AK002165.1 | - | 2 - 3 | 354 | 98 | ||
AK002180.1 | - | 2 - 3 | 359 | 98 | ||
AK222654.1 | - | 2 - 3 | 341 | 98 | ||
AK316585.1 | - | 2 - 3 | 345 | 98 | ||
AL080122.1 | - | 2 - 3 | 354 | 98 | ||
BC001553.1 | - | 2 - 3 | 197 | 3460712 | 98 | |
BI821848.1 | - | 2 - 3 | 358 | 98 | ||
BM828410.1 | - | 2 - 3 | 381 | 98 | ||
CR533456.1 | - | 2 - 3 | 121 | 98 | ||
DA426159.1 | - | 2 - 3 | 352 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000461.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell separation after cytokinesis
autophagy
nucleus organization
endosome organization
mitotic metaphase plate congression
regulation of centrosome duplication
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
cognition
neuron cellular homeostasis
multivesicular body sorting pathway
cell-cell adhesion
regulation of mitotic spindle assembly
positive regulation of viral release from host cell
ESCRT III complex disassembly
protein binding
protein domain specific binding
cadherin binding involved in cell-cell adhesion
cell separation after cytokinesis
autophagy
nucleus organization
endosome organization
mitotic metaphase plate congression
regulation of centrosome duplication
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
cognition
neuron cellular homeostasis
multivesicular body sorting pathway
cell-cell adhesion
regulation of mitotic spindle assembly
positive regulation of viral release from host cell
ESCRT III complex disassembly
protein binding
protein domain specific binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | TC03000461.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | ENST00000263780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | ENST00000471660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | ENST00000472024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | ENST00000494980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009019.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | TC03000461.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | ENST00000263780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | ENST00000471660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | ENST00000472024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009020.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157183.hg.1 | TC0300008002.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000466696.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | ENST00000494980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008002.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000263780.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000471660.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | ENST00000472024.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | NM_001244644.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | NM_014043.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157184.hg.1 | TC0300008002.hg.1 | Clariom D TC |
