Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023261.hg.1 |
TC04001669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023261.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023261.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183874.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183874.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183874.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183874.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183874.hg.1 |
TC0400012212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |