Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
ENST00000268852.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
ENST00000415126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
ENST00000431794.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
NM_001163735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
NM_025109.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018522.hg.1 |
TC17001402.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018523.hg.1 |
ENST00000268852.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018523.hg.1 |
ENST00000431794.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018523.hg.1 |
NM_001163735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018523.hg.1 |
NM_025109.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018523.hg.1 |
TC17001402.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000610930.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000610992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000611622.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000612392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000614623.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000620567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000621550.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
NM_001163735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
NM_025109.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000610930.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000610992.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000611622.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000612392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000614623.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000620567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
ENST00000621550.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
NM_001163735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
NM_025109.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206084.hg.1 |
TC1700012391.hg.1 |
Clariom D TC |