Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
TC02004954.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
ENST00000342045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
ENST00000344773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
ENST00000361704.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
ENST00000379624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
ENST00000379632.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
NM_004899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
NM_199191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
NM_199192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
NM_199193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002528.hg.1 |
NM_199194.2 |
Refseq |