Assay Details
| Gene Symbol: IAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017019253.1 | XP_016874742.1 | 1 - 2 | 88 | 75 | |
| GenBank mRNA | AA429852.1 | - | 2 - 3 | 75 | ||
| AA446156.1 | - | 1 - 2 | 55 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019253.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction
cell-cell signaling
sensory perception of pain
eating behavior
cellular protein metabolic process
negative regulation of cell differentiation
negative regulation of bone resorption
receptor binding
hormone activity
identical protein binding
apoptotic process
signal transduction
cell-cell signaling
sensory perception of pain
eating behavior
cellular protein metabolic process
negative regulation of cell differentiation
negative regulation of bone resorption
receptor binding
hormone activity
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000240652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000535428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000537593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000539393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | ENST00000542023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007050.hg.1 | NM_000415.2 | Refseq |

IAPP
Genomic Map