| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000261991.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000418736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000536315.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000554206.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000556594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
ENST00000614987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_004755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009973.hg.1 |
NM_182398.2 |
Refseq |