Assay Details
Gene Symbol: SUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080675.3 | NP_542406.2 | 1 - 2 | 168 | 85 | |
XM_011528573.1 | XP_011526875.1 | 1 - 2 | 183 | 85 | ||
XM_011528574.1 | XP_011526876.1 | 1 - 2 | 183 | 85 | ||
XM_011528576.1 | XP_011526878.1 | 1 - 2 | 184 | 85 | ||
GenBank mRNA | AF401350.1 | - | 1 - 2 | 168 | 85 | |
BC026118.2 | - | 1 - 2 | 168 | 4824412 | 85 | |
BC029528.1 | - | 1 - 2 | 161 | 5271065 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000767.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20010228.hg.1 | ENST00000356173.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010228.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010228.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010228.hg.1 | TC20000767.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010232.hg.1 | ENST00000356173.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010232.hg.1 | ENST00000375523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010232.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010232.hg.1 | TC20000767.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20010229.hg.1 | ENST00000356173.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010229.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010229.hg.1 | ENST00000375523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010229.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010229.hg.1 | TC20000767.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20010230.hg.1 | ENST00000356173.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010230.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010230.hg.1 | ENST00000375523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010230.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010230.hg.1 | TC20000767.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20010231.hg.1 | ENST00000356173.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010231.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010231.hg.1 | ENST00000375523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010231.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010231.hg.1 | TC20000767.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | TC2000008870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161807.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | TC2000008870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161813.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | TC2000008870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161809.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | TC2000008870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161810.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | TC2000008870.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161811.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000356173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000375523.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | ENST00000420875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008870.hg.1 | NM_080675.3 | Refseq |
