Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000321233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000341711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000342555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000394670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000394673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
ENST00000394679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
NM_001032293.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
NM_001098507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
NM_003457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004760.hg.1 |
TC17000375.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000321233.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000342555.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000394670.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000394673.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000577908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000581531.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000584696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_001032293.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_001098507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_003457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000321233.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000342555.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000394670.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000394673.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000577908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000581531.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
ENST00000584696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_001032293.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_001098507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
NM_003457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201831.hg.1 |
TC1700012230.hg.1 |
Clariom D TC |