Assay Details
| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011545566.2 | XP_011543868.1 | 1 - 2 | 103 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011545566.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000359680.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000380579.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000458511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000517553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000518847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | ENST00000519885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001011658.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_001128835.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009100.hg.1 | NM_014563.5 | Refseq |

TRAPPC2
Genomic Map