Assay Details
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142364.1 | NP_001135836.1 | 4 - 5 | 413 | 89 | |
NM_013338.4 | NP_037470.1 | 5 - 6 | 503 | 89 | ||
XM_017020552.1 | XP_016876041.1 | 4 - 5 | 441 | 89 | ||
GenBank mRNA | AF102850.1 | - | 5 - 6 | 511 | 89 | |
AF161498.1 | - | 5 - 6 | 452 | 89 | ||
AF183413.1 | - | 5 - 6 | 462 | 89 | ||
AK299085.1 | - | 4 - 5 | 389 | 89 | ||
AK299126.1 | - | 5 - 6 | 472 | 89 | ||
BC012531.1 | - | 5 - 6 | 455 | 4273297 | 89 | |
CB988638.1 | - | 500 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000570.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
protein N-linked glycosylation
GPI anchor biosynthetic process
determination of left/right symmetry
protein N-linked glycosylation via asparagine
dolichol metabolic process
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate beta-glucosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
protein glycosylation
protein N-linked glycosylation
GPI anchor biosynthetic process
determination of left/right symmetry
protein N-linked glycosylation via asparagine
dolichol metabolic process
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate beta-glucosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | ENST00000413537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | ENST00000443765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006480.hg.1 | TC13000570.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000460230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000486410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008632.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000239891.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000443765.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | ENST00000496689.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | NM_001142364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | NM_013338.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158240.hg.1 | TC1300008632.hg.1 | Clariom D TC |
