Assay Details
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246818.1 | XP_005246875.1 | 1 - 2 | 294 | 119 | |
GenBank mRNA | AF021834.1 | - | 1 - 2 | 238 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF021834.1, XM_005246818.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000412276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000428329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010223.hg.1 | ENST00000453517.1 | Ensembl |
