Assay Details
| Gene Symbol: PRMT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282956.1 | NP_001269885.1 | 3 - 4 | 629 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
DNA-templated transcription, termination
regulation of transcription, DNA-templated
regulation of mitotic nuclear division
cell proliferation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
circadian regulation of gene expression
peptidyl-arginine N-methylation
endothelial cell activation
histone H4-R3 methylation
regulation of DNA methylation
negative regulation of cell differentiation
positive regulation of oligodendrocyte differentiation
regulation of ERK1 and ERK2 cascade
Golgi ribbon formation
liver regeneration
regulation of signal transduction by p53 class mediator
negative regulation of nucleic acid-templated transcription
positive regulation of adenylate cyclase-inhibiting dopamine receptor signaling pathway
core promoter sequence-specific DNA binding
transcription corepressor activity
protein binding
methyltransferase activity
methyl-CpG binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
protein complex binding
protein-arginine omega-N symmetric methyltransferase activity
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)
protein heterodimerization activity
spliceosomal snRNP assembly
DNA-templated transcription, termination
regulation of transcription, DNA-templated
regulation of mitotic nuclear division
cell proliferation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
circadian regulation of gene expression
peptidyl-arginine N-methylation
endothelial cell activation
histone H4-R3 methylation
regulation of DNA methylation
negative regulation of cell differentiation
positive regulation of oligodendrocyte differentiation
regulation of ERK1 and ERK2 cascade
Golgi ribbon formation
liver regeneration
regulation of signal transduction by p53 class mediator
negative regulation of nucleic acid-templated transcription
positive regulation of adenylate cyclase-inhibiting dopamine receptor signaling pathway
core promoter sequence-specific DNA binding
transcription corepressor activity
protein binding
methyltransferase activity
methyl-CpG binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
protein complex binding
protein-arginine omega-N symmetric methyltransferase activity
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000216350.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000324366.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000397440.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000397441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000554716.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000555530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000556043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001039619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282953.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282955.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_006109.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000216350.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000324366.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000397440.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000397441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000553915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000554716.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000555530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | ENST00000556043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001039619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282953.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282955.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_001282956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | NM_006109.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162883.hg.1 | TC1400008684.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000216350.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000324366.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000397440.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000397441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000554716.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000555530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000556043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001039619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282953.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282955.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_006109.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000216350.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000324366.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000397440.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000397441.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553897.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000553915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000554716.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000555530.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | ENST00000556043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001039619.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282953.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282955.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_001282956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | NM_006109.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162882.hg.1 | TC1400008684.hg.1 | Clariom D TC |

PRMT5
Genomic Map