Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000477964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000507672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000508356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000508356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000508356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000508356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000508356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000509747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000509747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000509747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000509747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000512543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000512543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000512543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000512543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
ENST00000513787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013269.hg.1 |
NR_038402.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000477964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000506096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000511819.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013274.hg.1 |
ENST00000515878.1 |
Ensembl |