Assay Details
| Gene Symbol: RAB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163380.1 | NP_001156852.1 | 2 - 3 | 214 | 86 | |
| NM_032846.3 | NP_116235.2 | 3 - 4 | 287 | 86 | ||
| GenBank mRNA | AF468652.1 | - | 3 - 4 | 280 | 86 | |
| AK300599.1 | - | 2 - 3 | 183 | 86 | ||
| AK315354.1 | - | 3 - 4 | 267 | 86 | ||
| BC020839.1 | - | 3 - 4 | 276 | 4306410 | 86 | |
| BG535306.1 | - | 2 - 3 | 216 | 86 | ||
| BX647601.1 | - | 3 - 4 | 248 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000921.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300599.1,NM_001163380.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | ENST00000304034.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | ENST00000397762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010061.hg.1 | TC14000921.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000417141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000475857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000485996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008635.hg.1 | NR_028074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000397762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000461909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | ENST00000622293.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | NM_001163380.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | NM_032846.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162433.hg.1 | TC1400008635.hg.1 | Clariom D TC |

RAB2B
Genomic Map