Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002097.2 | NP_002088.2 | 5 - 6 | 757 | 96 | |
GenBank mRNA | AK057993.1 | - | 5 - 6 | 1293 | 96 | |
D32257.1 | - | 5 - 6 | 756 | 96 | ||
U20272.1 | - | 5 - 6 | 557 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000077.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13000794.hg.1 | ENST00000381140.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000794.hg.1 | ENST00000470606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000794.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000794.hg.1 | TC13000077.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13000795.hg.1 | ENST00000381140.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000795.hg.1 | ENST00000470606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13000795.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13000795.hg.1 | TC13000077.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000435781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000438571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000466776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000482655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006683.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000381140.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000419181.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000439403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000470606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | ENST00000493903.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | NM_002097.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300145603.hg.1 | TC1300006683.hg.1 | Clariom D TC |
