Assay Details
| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018942.2 | NP_061815.2 | 2 | 1830 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001021.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
multicellular organism development
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
multicellular organism development
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AF021232.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AF021233.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AF023849.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AF029761.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AK313528.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | AY358285.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | BC052270.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | BC077724.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 156211 | NM_003840.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 34638 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | chr4_random | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 40956 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3279009 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3328450 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2728 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P84320 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2727 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 34418 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46420 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53948 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3328450 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3279009 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00319890 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3279009 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3328450 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P84320 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P03808674 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03808682 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03808674 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P41907569 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P84320 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 36949 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03808674 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P24086314 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P24086507 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41907555 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03808674 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P24086507 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_018942_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8104137 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8104138 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8104136 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207353_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11733657_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11733657_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014069.hg.1 | TC04001021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014069.hg.1 | ENST00000400677.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014069.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000506970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000506970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168651.hg.1 | TC0400009992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000400677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000506970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000506970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | ENST00000617742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_001306142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009992.hg.1 | NM_018942.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 214386_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214386_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207353_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207353_s_at | - | - |
| Human Genome U133A 2.0 Array | 214386_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214386_at | - | - |
| Human Genome U133A 2.0 Array | 207353_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 207353_s_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_018942_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw500557 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw581971 | - | - |
| Human Genome U95 Set | 39535_PM_at | - | - |
| Human Genome U95 Set | 39535_at | - | - |
| Human Genome U95 Set | 39536_PM_at | - | - |
| Human Genome U95 Set | 39536_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207353_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214386_at4 | - | - |
| Human Genome U133 Set | 207353_PM_s_at | - | - |
| Human Genome U133 Set | 207353_s_at | - | - |
| Human Genome U133 Set | 214386_PM_at | - | - |
| Human Genome U133 Set | 214386_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023928 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1796376 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_9506784-S | M99587.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_22747-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0001400017 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0001400017 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1796376 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1796376 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5690113 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_22747 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1811022 | M99587.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1796376 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1811022 | M99587.1 | human_transcriptome |

HMX1
Genomic Map