Assay Details
| Gene Symbol: GRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017551.2 | NP_060021.1 | 11 - 12 | 1957 | 68 | |
| XM_011539720.2 | XP_011538022.1 | 11 - 12 | 1954 | 68 | ||
| GenBank mRNA | AB033046.1 | - | 8 - 9 | 1204 | 68 | |
| AK127168.1 | - | 11 - 12 | 1942 | 68 | ||
| AK293104.1 | - | 11 - 12 | 1915 | 68 | ||
| AK302192.1 | - | 4 - 5 | 729 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018672.hg.1 | ENST00000327946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018672.hg.1 | ENST00000536331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018672.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018672.hg.1 | TC10001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176777.hg.1 | TC1000011287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |

GRID1
Genomic Map