Assay Details
Gene Symbol: TUBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193609.1 | NP_001180538.1 | 5 - 6 | 1053 | 127 | |
NM_001193612.1 | NP_001180541.1 | 6 - 7 | 1149 | 127 | ||
XM_017024719.1 | XP_016880208.1 | 4 - 5 | 839 | 127 | ||
XM_017024720.1 | XP_016880209.1 | 3 - 4 | 628 | 127 | ||
GenBank mRNA | AB031044.1 | - | 4 - 5 | 766 | 127 | |
AK315172.1 | - | 6 - 7 | 1145 | 127 | ||
BC000258.1 | - | 5 - 6 | 1028 | 3358263 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001753.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
