Assay Details
| Gene Symbol: AATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012138.3 | NP_036270.1 | 2 - 3 | 534 | 67 | |
| XM_011524611.1 | XP_011522913.1 | 2 - 3 | 518 | 67 | ||
| GenBank mRNA | AF083208.1 | - | 2 - 3 | 451 | 67 | |
| AJ249940.2 | - | 2 - 3 | 389 | 67 | ||
| AK057229.1 | - | 2 - 3 | 534 | 67 | ||
| BC000591.2 | - | 2 - 3 | 414 | 3346769 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000444.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to DNA damage stimulus
cell adhesion
regulation of mitotic cell cycle
negative regulation of superoxide anion generation
embryonic cleavage
ribosome biogenesis
negative regulation of amyloid precursor protein biosynthetic process
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of reactive oxygen species metabolic process
transcription factor activity, sequence-specific DNA binding
protein binding
leucine zipper domain binding
poly(A) RNA binding
tau protein binding
cellular response to DNA damage stimulus
cell adhesion
regulation of mitotic cell cycle
negative regulation of superoxide anion generation
embryonic cleavage
ribosome biogenesis
negative regulation of amyloid precursor protein biosynthetic process
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of reactive oxygen species metabolic process
transcription factor activity, sequence-specific DNA binding
protein binding
leucine zipper domain binding
poly(A) RNA binding
tau protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005262.hg.1 | ENST00000225402.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005262.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005262.hg.1 | TC17000444.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700155238.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155238.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155238.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155238.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155238.hg.1 | TC1700007658.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000610798.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000613840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000615319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616392.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000616434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000617141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000619387.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | ENST00000622432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007658.hg.1 | NM_012138.3 | Refseq |

AATF
Genomic Map