Assay Details
| Gene Symbol: TCEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003198.2 | NP_003189.2 | 1 - 2 | 216 | 96 | |
| GenBank mRNA | AF116729.1 | - | 1 - 2 | 65 | 96 | |
| AK310229.1 | - | 1 - 2 | 202 | 96 | ||
| AK313079.1 | - | 1 - 2 | 88 | 96 | ||
| BC002883.2 | - | 1 - 2 | 100 | 3941728 | 96 | |
| DB461774.1 | - | 1 - 2 | 254 | 96 | ||
| L47345.1 | - | 1 - 2 | 110 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000300.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF116729.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004549.hg.1 | TC01000300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004549.hg.1 | ENST00000418390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004549.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004548.hg.1 | TC01000300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004548.hg.1 | ENST00000418390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004548.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004551.hg.1 | TC01000300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004551.hg.1 | ENST00000418390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004551.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004551.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151029.hg.1 | TC0100007331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151032.hg.1 | TC0100007331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151030.hg.1 | TC0100007331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151034.hg.1 | TC0100007331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 203758 | - | - |

TCEB3
Genomic Map