Assay Details
| Gene Symbol: WDR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522699.2 | XP_011521001.1 | 2 - 3 | 1773 | 87 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |

WDR24
Genomic Map