Assay Details
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002538.3 | NP_002529.1 | 1 - 2 | 365 | 96 | |
XM_017008914.1 | XP_016864403.1 | 1 - 2 | 110 | 96 | ||
GenBank mRNA | U49184.1 | - | 1 - 2 | 96 | 96 | |
U53823.1 | - | 1 - 2 | 366 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000304.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_002538.3,U49184.1,U53823.1,XM_017008914.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004072.hg.1 | TC05000304.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05004072.hg.1 | ENST00000355237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004072.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | TC05000304.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | ENST00000355237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | ENST00000380766.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | ENST00000396442.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | ENST00000538151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | ENST00000542132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004077.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152091.hg.1 | TC0500007681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500152087.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152087.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152087.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152087.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152087.hg.1 | TC0500007681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 108955 | - | - |
