Assay Details
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205254.1 | NP_001192183.1 | 3 - 4 | 940 | 98 | |
NM_002538.3 | NP_002529.1 | 3 - 4 | 1170 | 98 | ||
GenBank mRNA | AB451306.1 | - | 2 - 3 | 734 | 98 | |
AB451437.1 | - | 2 - 3 | 734 | 98 | ||
AK290697.1 | - | 3 - 4 | 902 | 98 | ||
BC029886.1 | - | 3 - 4 | 912 | 5179203 | 98 | |
GQ402517.1 | - | 2 - 3 | 778 | 98 | ||
U49184.1 | - | 3 - 4 | 901 | 98 | ||
U53823.1 | - | 3 - 4 | 1171 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000304.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | TC05000304.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | ENST00000355237.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | ENST00000396442.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | ENST00000538151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004080.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152095.hg.1 | TC0500007681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000355237.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000396442.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000510666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000514370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | ENST00000538151.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205254.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_001205255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007681.hg.1 | NM_002538.3 | Refseq |
