Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
TC05000280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000284037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000380935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000380936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000380938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000380939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000380943.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000506030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000508515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
ENST00000511297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_001006600.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_001253698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_001253699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_001253701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003573.hg.1 |
NM_018695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003576.hg.1 |
TC05000280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003576.hg.1 |
ENST00000284037.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003576.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000284037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000511671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000512354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000284037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000511671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
ENST00000512354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151433.hg.1 |
TC0500007610.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000284037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380938.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000506030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000508515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000511297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000511671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001006600.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_018695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000284037.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380938.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000380943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000506030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000508515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000511297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
ENST00000511671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001006600.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253697.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_001253701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
NM_018695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151430.hg.1 |
TC0500007610.hg.1 |
Clariom D TC |