| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000256733.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000256733.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000256733.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000256733.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000256733.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000278222.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000363466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000363466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000414546.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000414546.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000414546.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000414546.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000524555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000526900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000528349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000528349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000528349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000528349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000529528.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000530400.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000530400.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
ENST00000530400.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001127380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_001199744.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_006512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013152.hg.1 |
NM_030754.4 |
Refseq |