Assay Details
| Gene Symbol: ARL6IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313858.1 | NP_001300787.1 | 2 - 3 | 299 | 69 | |
| NM_015161.2 | NP_055976.1 | 2 - 3 | 375 | 69 | ||
| GenBank mRNA | AK291137.1 | - | 2 - 3 | 241 | 69 | |
| AK300825.1 | - | 3 - 4 | 183 | 69 | ||
| AK300869.1 | - | 2 - 3 | 244 | 69 | ||
| AK301636.1 | - | 2 - 3 | 299 | 69 | ||
| AK307315.1 | - | 2 - 3 | 244 | 69 | ||
| BC010281.1 | - | 2 - 3 | 232 | 3048642 | 69 | |
| D31885.1 | - | 2 - 3 | 232 | 69 | ||
| DC406882.1 | - | 2 - 3 | 377 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000915.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300825.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of L-glutamate transport
cotranslational protein targeting to membrane
apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
endoplasmic reticulum tubular network assembly
endoplasmic reticulum tubular network membrane organization
protein binding
positive regulation of L-glutamate transport
cotranslational protein targeting to membrane
apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
endoplasmic reticulum tubular network assembly
endoplasmic reticulum tubular network membrane organization
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16013112.hg.1 | ENST00000304414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013112.hg.1 | ENST00000545430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013112.hg.1 | ENST00000546206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013112.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013112.hg.1 | TC16000915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013111.hg.1 | ENST00000304414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013111.hg.1 | ENST00000545430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013111.hg.1 | ENST00000546206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013111.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013111.hg.1 | TC16000915.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192938.hg.1 | TC1600011494.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192934.hg.1 | TC1600011494.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192930.hg.1 | TC1600011494.hg.1 | Clariom D TC |

ARL6IP1
Genomic Map