Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006526.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144626.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144626.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144626.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144626.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144626.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144620.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144620.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144620.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144620.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144620.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144619.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144619.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144619.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144622.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144622.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144622.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144622.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144622.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144624.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144624.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144624.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144624.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144624.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144621.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144621.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144621.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144621.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144621.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144623.hg.1 |
TC2200006526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144623.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144623.hg.1 |
NR_103718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144623.hg.1 |
ENST00000600723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144623.hg.1 |
NR_103718.1 |
Refseq |