Assay Details
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011514841.1 | XP_011513143.1 | 1 - 2 | 423 | 64 | |
XM_011514842.1 | XP_011513144.1 | 1 - 2 | 423 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011514841.1, XM_011514842.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000339139.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000616725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | ENST00000618917.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142407.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142408.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142417.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001142435.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_001261822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012002.hg.1 | NM_003296.3 | Refseq |
