Assay Details
| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020445.5 | NP_065178.1 | 10 - 11 | 1208 | 115 | |
| NR_073000.1 | 9 - 10 | 1152 | 115 | |||
| NR_073001.1 | 9 - 10 | 1083 | 115 | |||
| XM_017012462.1 | XP_016867951.1 | 10 - 11 | 1119 | 115 | ||
| XM_017012463.1 | XP_016867952.1 | 10 - 11 | 1058 | 115 | ||
| GenBank mRNA | AB209174.1 | - | 6 - 7 | 4535 | 115 | |
| AF023453.1 | - | 10 - 11 | 1110 | 115 | ||
| AK001198.1 | - | 6 - 7 | 676 | 115 | ||
| AK294292.1 | - | 9 - 10 | 1106 | 115 | ||
| AK315534.1 | - | 10 - 11 | 1190 | 115 | ||
| AK316078.1 | - | 9 - 10 | 1003 | 115 | ||
| BC008682.1 | - | 10 - 11 | 1163 | 3854008 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001031.hg.1
Set Membership: |
|

ACTR3B
Genomic Map