Assay Details
| Gene Symbol: DCAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005828.4 | NP_005819.3 | 5 - 6 | 982 | 80 | |
| NR_073585.1 | 5 - 6 | 982 | 80 | |||
| GenBank mRNA | BC001264.1 | - | 5 - 6 | 935 | 3458293 | 80 |
| U94747.1 | - | 5 - 6 | 811 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000768.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17010000.hg.1 | ENST00000310827.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010000.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010000.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010000.hg.1 | TC17000768.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163987.hg.1 | TC1700008563.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000580091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000582274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | ENST00000615512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008563.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000415273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000431926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | ENST00000614556.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | NM_005828.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | NR_073585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700163988.hg.1 | TC1700008563.hg.1 | Clariom D TC |

DCAF7
Genomic Map