Assay Details
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011529441.1 | XP_011527743.1 | 1 - 2 | 245 | 85 | |
XM_017028265.1 | XP_016883754.1 | 1 - 2 | 195 | 85 | ||
XM_017028266.1 | XP_016883755.1 | 1 - 2 | 195 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529441.1, XM_017028265.1, XM_017028266.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000626994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006663.hg.1 | ENST00000630155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
