Assay Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028265.1 | XP_016883754.1 | 2 - 3 | 385 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028265.1 transcripts.
Set Membership: |
|

BTG3
Genomic Map